Learn More
anti-RASGRP3, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00025780-PW1.1Set
Product Code. 16043549
  • 809.00€ / 1 set
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Antibody pair for IP: rabbit polyclonal anti-RASGRP3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-RASGRP3 (50μg)
Specifications
RASGRP3
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
GRP3/KIAA0846
Mouse, Rabbit
RUO
25780
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
RASGRP3
RASGRP3
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-RASGRP3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-RASGRP3 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.