missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RBP4 Matched Antibody Pair, Abnova
Brand: Abnova H00005950-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-RBP4 (100μg); Mouse polyclonal anti-RBP4 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells.
ELISA
Specifications
RBP4 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RBP4. | |
RBP4 | |
Affinity Purified | |
RUO | |
5950 | |
Human | |
Serum |
ELISA | |
Unconjugated | |
RBP4 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RBP4 (100 μg) 2. Detection antibody: mouse polyclonal anti-RBP4 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |