Learn More
anti-RNF32, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00140545-PW3.1Set
Product Code. 16153864
  • 809.00€ / 1 set
Includes: Antibody pair for IP: rabbit purified polyclonal anti-RNF32 (300ÎĽg), Antibody pair for WB: mouse purified polyclonal anti-RNF32 (50ÎĽg)
Specifications
RNF32
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FKSG33/HSD15
Mouse, Rabbit
RUO
140545
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
RNF32
RNF32
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit purified polyclonal anti-RNF32 (300 ug) 2. Antibody pair for WB: mouse purified polyclonal anti-RNF32 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
anti-RNF32, Human, IP-WB Antibody Pair, Abnova™ > 1 Set
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado
For Research Use Only