missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUN and FYVE domain containing 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RUFY1.
Brand: Abnova H00080230-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: ITSFEGKTNQVMSSMKQMEERLQHSERARQGAEERSHKLQQELGGRIGALQLQLSQLHEQCSSLEKELKSEKEQRQALQRELQHEKDTSSLLRMELQQVSpecifications
RUN and FYVE domain containing 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant RUFY1. | |
RUFY1 | |
FLJ22251/RABIP4/ZFYVE12 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_025158 | |
RUFY1 | |
RUFY1 (NP_079434, 376 a.a. ∼ 474 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
80230 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |