missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUN domain containing 2A, Mouse, Clone: 6D6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant RUNDC2A.
Brand: Abnova H00084127-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MDEERSSMLPTMAAGPNSILFAINIDNKDLNGQSKFAPTVSDLLKESTQNVTLLKESTQGVSSVFREITASSAISILIKPEQETDPLPVVSRNVSADAKCKKERKKKKQVTNIISFDDEEDEQNSGDMFKKTPGAGESSEDNSDHSSVNIMSAFESPFGPNSNGSQSSNSWKIDSLSLNREFGYQKLDVKSIDDEDVDENEDDVYGNSSGRKHRGHSESPEKNGAHSVTQAGVQWHDLGSLQPLPPGFRSpecifications
RUN domain containing 2A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC041583 | |
RUNDC2A | |
RUNDC2A (AAH41583, 1 a.a. ∼ 249 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
84127 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunofluorescence, Western Blot | |
6D6 | |
Mouse monoclonal antibody raised against a full length recombinant RUNDC2A. | |
RUNDC2A | |
FLJ12363 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
Antibody |