missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPD, Human, Matched Antibody Pair (AP42), Abnova™
Brand: Abnova H00006441-AP42.1Set
Includes: Capture antibody: mouse monoclonal anti-SFTPD (100μg); Detection antibody: biotinylated mouse monoclonal anti-SFTPD, IgG2a Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SFTPD.
Host Species: Mouse, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein),ELISA Pair (Transfected Lysate)
Specifications
SFTPD | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SFTPD. | |
COLEC7/PSP-D/SFTP4/SP-D | |
Mouse | |
RUO | |
6441 | |
Human |
ELISA | |
Unconjugated | |
SFTPD | |
SFTPD | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-SFTPD (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-SFTPD, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SFTPD, Human, Matched Antibody Pair (AP42), Abnova™ > 1 set, Unlabeled, Biotinylated
Spot an opportunity for improvement?Share a Content Correction