missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-STX1A Matched Antibody Pair, Abnova
Brand: Abnova H00006804-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-STX1A (100μg); Mouse polyclonal anti-STX1A (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Synaptic vesicles store neurotransmitters that are released during calcium-regulated exocytosis. The specificity of neurotransmitter release requires the localization of both synaptic vesicles and calcium channels to the presynaptic active zone. Syntaxins function in this vesicle fusion process. Syntaxins also serve as a substrate for botulinum neurotoxin type C, a metalloprotease that blocks exocytosis and has high affinity for a molecular complex that includes the α-latrotoxin receptor (MIM 600565) which produces explosive exocytosis.
ELISA
Specifications
STX1A | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human STX1A. | |
HPC-1/STX1/p35-1 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-STX1A (100 μg) 2. Detection antibody: mouse polyclonal anti-STX1A (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
STX1A | |
STX1A | |
Affinity Purified | |
RUO | |
6804 | |
Human | |
Serum |