Learn More
TNFR Related Protein Mouse anti-Human, PE, Clone: HTNFR-RP-M12, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551503
463.62 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The hTNFR-RP-M12 antibody reacts with the extracellular domain of the 61kDa receptor for the human cytokines, LTα1β2 and LIGHT. This receptor is referred as the Lymphotoxin β Receptor (LTβR). The gene encoding LTβR has been designated as the TNFRSF3 by the Human Gene Nomenculture Committee. LTβR was previously known as TNFRIII and TNF receptor-related protein (TNFRrp). LTβR is a type I transmembrane glycoprotein and member of the TNF Receptor Superfamily. LTβR are expressed on stromal cells in lymphoid tissue, normal dermal fibroblasts, bronchial airway epithelial cells and in a variety of adherent cell lines including FDC-1, U937, HT-29, HeLa and HEK 293 cells. LTβR are absent on human peripheral blood T and B cells and expressed at low levels by monocytes.
Host Species: Mouse
Clone: hTNFR-RP-M12
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human LTβR-Fc protein
Flow Cytometry

Specifications
TNFR Related Protein | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human LTβR-Fc protein | |
0.2 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
HTNFR-RP-M12 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.