missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-3 Tubulin Monoclonal Antibody (2E9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515541
This item is not returnable.
View return policy
Description
MA5-15541 targets TUBB3 in indirect ELISA, FACS, IF, and WB applications and shows reactivity with Human samples. The MA5-15541 immunogen is purified recombinant fragment of human TUBB3 expressed in E. Coli. MA5-15541 detects TUBB3 which has a predicted molecular weight of approximately 50kDa.
The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.Specifications
beta-3 Tubulin | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q13509 | |
TUBB3 | |
Purified recombinant fragment of human TUBB3 expressed in E. Coli. | |
RUO | |
10381 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
2E9 | |
Unconjugated | |
TUBB3 | |
3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115) | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |