missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-UBL4A Matched Antibody Pair, Abnova™
Brand: Abnova H00008266-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-UBL4A (100μg); Detection antibody: mouse purified polyclonal anti-UBL4A (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human UBL4A.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
UBL4A | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human UBL4A. | |
DX254E/DXS254E/G6PD/GDX/UBL4 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-UBL4A (100 μg) 2. Detection antibody: mouse purified polyclonal anti-UBL4A (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
UBL4A | |
UBL4A | |
Affinity Purified | |
RUO | |
8266 | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
anti-UBL4A Matched Antibody Pair, Abnova™ > 1 set, Unlabeled
Spot an opportunity for improvement?Share a Content Correction