Learn More
Macherey Nagel Aquadur™ Water Hardness Test Strips
For semi-quantitative determination of water hardness (CaC03)
Brand: Macherey Nagel 91224
This item is not returnable.
View return policy
Description
- Test papers allow the semi-qualitative determination of water hardness
- Ranged test results
Specifications
CaCO3 | |
1000 individually sealed strips | |
<54, >72, >26, >252, >375 ppm | |
Test strips, semi-quantitative, water hardness |