missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL6 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97182F
This item is not returnable.
View return policy
Description
ARL6 Polyclonal antibody specifically detects ARL6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
ARL6 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
FITC | |
ADP-ribosylation factor-like 6, ADP-ribosylation factor-like protein 6, Bardet-Biedl syndrome 3 protein, BBS3, MGC32934, RP55 | |
Produced in rabbits immunized with E. coli-derived Human ARL6 fragment. | |
0.1 mL | |
Signal Transduction | |
84100 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARL6 Antibody, FITC, Novus Biologicals™ > 0.1 mL, FITC
Spot an opportunity for improvement?Share a Content Correction