missing translation for 'onlineSavingsMsg'
Learn More

Arsenic AAS Standard, 1000 mg/L As in HNO3 2%, Honeywell Fluka™

Product Code. p-7236093 Shop All Honeywell Fluka Products
100 mL, 100mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic AAS Standard
Density 1,015 g/cm3
pH <2
Quantity 100 mL
Grade Standards and Modifiers for AAS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Arsenic AAS Standard, 1000 mg/L As in HNO3 2%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.