Learn More
Thermo Scientific™ ART™ Extended Length Tips
Superior tips for any application, any pipette
Brand: Thermo Scientific™ 3510
19.11 EUR valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Superior tips for any application, any pipette
Thermo Scientific™ ART™ Extended Length Tips deliver proven ART quality, performance and reliability,backed by the highest Thermo Scientific standards and worldwide support.
Every tip is certified to meet the industry's highest standards for molding precision and clarity, and are certified free of RNase, DNase, ATP, and Pyrogens. And each meets the exact specifications of its respective pipette brand manufacturer.
Specifications
ART 10 REACH, Non-Filtered Tip | |
Non-filtered | |
Bulk |
10 μL | |
Non-sterile | |
ART |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
13Hours
8Minutes
8Seconds
33New Offer Save Up to 92% -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
30Hours
8Minutes
8Seconds
33 -
Thermo Scientific ClipTip
,
Thermo Scientific ART
,
Thermo Scientific Finnpipette
,
Thermo Scientific Matrix
Exclusive Promotion on Liquid Handling
Unlock Precision: Top Pipettes and Tips on Promotion!Promo Code: 25076Expires: 2025-06-30
Days
91Hours
8Minutes
8Seconds
33 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
8Minutes
8Seconds
33