Learn More
Fisherbrand™ Grade 51 Ashless Hardened Quantitative Filter Paper
Used for quantitative analysis.
Brand:  Fisherbrand™ 0051A00009
 View more versions of this product
Product Code. 11542053
  • 22.60€ / Pack of 100
Estimated Shipment: 16-10-2024
Description
  • Plain disc, wet strengthened ashless filter paper
  • Made with 100% cotton linters fibers
  • Paper have been washed out with acid
  • Finally cleaned with demineralized water
  • Ash content: 0.007%
Applications:
  • Used for analysis of acid solution and pressure filtration
  • Food industry
  • Beverage industry
  • Atmospheric analysis
Specifications
Specifications
Ash Content: <0.008%
Cotton Linters
12 to 15 μm
Acid solutions analysis and for filtration under high pressure
100 Pack
150 mm
Disc
12 to 15 μm
10 sec.
51
0.18 mm