missing translation for 'onlineSavingsMsg'
Learn More

ATP Synthase B1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5106954

Product Code. 17146763

  • 526.00€ / 100µL
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene encodes the b subunit of the proton channel.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP Synthase B1
Polyclonal
Unconjugated
ATP5PB
ATP synthase B chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase peripheral stalk-membrane subunit b; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase subunit b; ATP synthase subunit b, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1; Atp5f; ATP5F1; Atp5pb; ATPase subunit b; C76477; cell proliferation-inducing protein 47; H+-ATP synthase subunit b; PIG47
Rabbit
Affinity Chromatography
RUO
515
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
P24539
ATP5PB
Synthetic peptide located within the following region: VTYRERLYRV YKEVKNRLDY HISVQNMMRR KEQEHMINWV EKHVVQSIST
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.