missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517265
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.Specifications
ATR | |
Polyclonal | |
Unconjugated | |
ATR | |
2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
545 | |
-20°C | |
Liquid |
Western Blot | |
14 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q13535 | |
ATR | |
Synthetic peptide corresponding to central residues of human ATR | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |