Learn More
ATR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA585507
Additional Details : Weight : 0.01000kg
Description
Keep as concentrated solution. Positive Control: 293T, A431, HeLa, HepG2, Raji, Raji NE, Raji nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.Specifications
ATR | |
Polyclonal | |
Unconjugated | |
ATR | |
2310008J16Rik; 2810405N18Rik; ataxia telangiectasia and Rad3 related; ataxia telangiectasia and Rad3-related protein; ATR; ATR serine/threonine kinase; EC 2.7.11.1; FCTCS; FRAP-related protein 1; FRAP-related protein-1; FRP1; I79_000761; kinase ATR; MEC1; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; SCKL; SCKL1; Serine/threonine-protein kinase ATR; TEM8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
245000, 545 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q13535, Q9JKK8 | |
ATR | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ATR. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |