missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit

Competitive ELISA

4730.00€

Specifications

Assay Sensitivity 0.25 ng/mL
Description Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA Kit
View More Specs

Products 1
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
17825117
View Documents
Invitrogen™
EIAANPX10
10 x 96 Assays
4730.00€
Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
 
Description

Description

The Atrial Natriuretic Peptide (ANP) Competitive ELISA quantitates ANP in human plasma,urine or cell culture medium.

Principle of the method

The Atrial Natriuretic Peptide (ANP) Competitive ELISA research-use-only kit is designed to quantitatively measure ANP present in plasma,urine,and tissue culture media samples. This kit is not recommended for serum or plasma samples without extraction. An ANP standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An ANP-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of an antibody to ANP to each well. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound ANP-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

ANP is derived from the 151 amino acid long protein preproANP3. The prepro protein is cleaved to generate proANP. ANP is derived from amino acids 99-126 to form the 28 amino acid peptide with a disulfide bond between amino acids 7 and 23. ANP is the predominant member of a family of structurally and functionally related peptide hormones that exert a wide array of effects on cardiovascular and renal function. ANP is identical in a large number of species. Human, pig, sheep, and dog ANP have identical sequences and this kit will measure samples from these species identically. Rat and mouse ANP molecules differ at amino acid 12 but show complete cross reactivity with this kit.
Specifications

Specifications

0.25 ng/mL
HRP
ELISA Kit
Human
Colorimetric Microplate Reader
6.7%
RUO
-20°C
1 hr. 20 min.
Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit
Fluorescence
Plasma, Supernatant, Urine
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
7.6%
HRP
Inflammation
Chemical
1 hr. 30 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.