missing translation for 'onlineSavingsMsg'
Learn More
Atropine sulfate monohydrate, 97+%, Thermo Scientific Chemicals Product Code.: 10464525

Atropine sulfate monohydrate, 97+%, Thermo Scientific Chemicals

Product Code. 10464525 Shop All Thermo Scientific Chemicals Products
100 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10464525

Brand: Thermo Scientific Acros 226681000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Atropine sulfate monohydrate, CAS # 5908-99-6, is a muscarinic receptor antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Atropine is a competitive and reversible antagonist of muscarinic acetylcholine receptors on both central and peripheral neurons
  • Atropine was originally isolated from the belladonna plant Atropa belladonna but is also found in other Solanaceae plants
  • Atropine is a drug that can be used to slow heart rate, decrease salivary secretions, and treat poisoning caused by cholinergic or muscarinic agents

Applications

  • It is used to block activation of muscarinic receptors in laboratory mice
  • It can be used in a laboratory setting to decrease oral and airway secretions in research animals
  • Atropine has been used to study heart rate and blood pressure variability in swine
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5908-99-6
Molecular Formula H2SO4·H2O
Molecular Weight (g/mol) 694.85
MDL Number MFCD00074815
InChI Key PVGPXGKNDGTPTD-IJTOKZDFSA-N
Synonym atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acidShow MoreShow Less
PubChem CID 23624044
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O

Specifications

Acidity Passes Test
Melting Point >187.0°C
Color White
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 100 g
Beilstein 21, 27
Merck Index 15, 866
Specific Rotation Condition −0.60° (25°C per USP)
Specific Rotation − 0.60
Solubility Information Solubility in water: soluble. Other solubilities: soluble in boiling alcohol and glycerol, slightly soluble in chloroform, practically insoluble in ether
Formula Weight 694.85
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Atropine sulfate monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471315 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number226680100,226681000,226680250
Lot NumberA0448731 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog Number226680100,226681000,226680250
Lot NumberA0444633 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number226680100,226681000,226680250
Lot NumberA0440787 Certificate TypeCertificate of Analysis Date06/04/2022 Catalog Number226680100,226681000,226680250
Lot NumberA0437082 Certificate TypeCertificate of Analysis Date17/12/2021 Catalog Number226680100,226681000,226680250
compliance-icons
Product Identifier
  • Atropine sulfate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Atropine sulfate monohydrate, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.