Learn More
3M™ Aura™ Particulate Respirators 9300+ Series
Particulate respirators 9300+ Series feature an ergonomic, flat-fold, 3-panel design with a low breathing resistance filter media. Available in FFP1, FFP2 and FFP3 protection levels with valved and unvalved models, respirators have an innovative chin tab for easy adjustments and an embossed, sculpted upper panel to help reduce eyewear fogging.
Brand: 3M™ 7100258910
33.89 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Particulate respirators 9300+ Series feature an ergonomic, flat-fold, 3-panel design with a low breathing resistance filter media. Available in FFP1, FFP2 and FFP3 protection levels with valved and unvalved models, respirators have an innovative chin tab for easy adjustments and an embossed, sculpted upper panel to help reduce eyewear fogging.
- Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
- Offers FFP1, FFP2 and FFP3 protection.
- 3-panel design allows for greater facial movement.
- Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
- Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
- Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
- Individually packed in hygienic wrapping to prevent contamination before use.
- Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
- Foldable design makes them easy to store when not in use.
- Respirators work well with 3M eyewear and hearing protection products.
Specifications
10 | |
EN 149 | |
Flat-fold | |
FFP2 | |
General Purpose Particulate Respirator | |
No | |
Yes | |
9320+ | |
Sculpted | |
Dusts and mists found in a wide variety of industrial applications and other work situations | |
Non-sterile | |
Colour-coded Headband, Blue |
European Standard EN 149 for Filtering Half Masks to Protect Against Particles | |
White | |
Single-use | |
Polypropylene | |
One Size | |
Polypropylene | |
Polyisoprene | |
Aluminum, Polyurethene | |
Individually Packaged | |
Cup | |
3-panel | |
No |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now