Learn More
Abeba™ Autoclavable Clogs 9620
Antistatic and autoclavable at 121°C. High slip resistance.
Brand: Abeba™ 9620/41
52.38 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- Green
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
CE EN ISO 20347:2012 OB A, E SRC | |
Green | |
41 | |
9620 |