Learn More
Raypa™ AE-Dry Autoclave
Floor-standing automatic vertical autoclave with top loading access. Raypa™ AE-Dry Autoclave delivers great performance for general labware sterilization in industry, education, medicine, and research.
Brand: Raypa™ AE-75-DRY
This item is not returnable.
View return policy
Description
- Intended for the sterilization of a wide range of solid materials such as unwrapped and wrapped items, small porous objects and hollow ware type B (slightly deep and broad cavities), glass, plastic, metallic objects and waste bags.
- The drying feature and the automatic water control level reduce operator time dedicated to the autoclave leading to facilitate the sterilization procedures in the general laboratory.
- Agar mode (40 to 80°C).
- Heart probe (optional).
- Programmable auto-start (24 hours).
- Heart probe (optional).
- Built in or benchtop printer (optional).
- Chamber, lid, closure, top and external case made of AISI-304 stainless steel.
- Total control by microprocessor: ten sterilization programs, six of them are user free.
- Feeding water reservoir.
- Automatic chamber filling.
- Automatic drying.
- RS232 connection for PC/printer.
- Inlet in chamber for an external validation probe.
- Supplied with a protecting grid for heater.
- Safety valve and thermostat.
- Hydraulic door blocking system while existing positive pressure in the chamber.
- Safety pressure switch.
- Open lid sensor.
Specifications
79 L | |
Autoclave, Top-loading | |
140 kg | |
73 x 62 x 119 cm Exterior |
40 x 60 cm Interior | |
100°C to 134°C | |
2.1 bar max. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Raypa™ AE-Dry Autoclave