Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
Brand:  BRAND™ 43088
101.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10012392
  • 130.00€ / Each
Only null left
Estimated Shipment: 17-06-2025
Description
  • Dispenser head
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass 3.3
Without bottle
100 mL
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Dispenser, tilt measure without bottle > Capacity: 100 mL