Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
Brand: BRAND™ 43078
82.83 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Calibrated to deliver (TD, Ex)
- Inscriptions in green enamel
- The automatic dispenser is particularly well-suited for dispensing suspensions
Specifications
Borosilicate Glass | |
Without bottle | |
50 mL |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
1Hours
18Minutes
9Seconds
32
Product Content Correction
Product Title
BRAND™ Dispenser, tilt measure without bottle > Capacity (Metric): 50 mL