Learn More
Carbolite™ AX Drying Oven
Bench-top sized laboratory oven. Carbolite™ AX Drying Oven is equipped with the R38 digital PID temperature controller.
Brand:  Carbolite™ AX030-230SN+&02-R38
 View more versions of this product
Product Code. 12029802
  • 3410.00€ / Each
Only null left
Estimated Shipment: 08-08-2025
Description
  • 250°C maximum operating temperature
  • 30, or 60L chamber volumes
  • Equipped with the R38 digital PID temperature controller
  • Fan convection for rapid heating and excellent uniformity
  • Chemically resistant stainless steel liner
  • Two adjustable nickel-chrome plated wire shelves
  • Lever latch door and airtight silicone seal
  • Compliant with safety standard BS EN 61010-2-010:2003
  • Options available (specify at time of order)
    • Over-temperature protection (recommended to protect valuable contents and for unattended operation)
    • Digital countdown timer to switch oven off
    • Additional sets of shelves and runners
    • Lockable door
    • Low voltage options for use below 220 V
    • Routine spares kit
Specifications
Specifications
BS EN 61010-2-010:2003
Ambient +30° to 250°C
Digital
Stainless Steel
AX30
±5°C
590 x 465 x 440 mm
465 mm
28 L
Drying Oven
Low thermal mass insulation
10 kg/shelf
2 (Fitted), 4 (Accepted)
295 x 300 x 320 mm
590 mm
440 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Carbolite™ AX Drying Oven > Chamber Volume: 28L