Learn More
Axygen™ Tip Refill System
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
Brand: Axygen™ RFL-222-C
37.75 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
Specifications
1 to 200 μL | |
Clear | |
Universal | |
Yes |
Non-sterile | |
Autoclavable | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%