Learn More
Axygen™ Axypet™ Single-Channel Pipettor
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
50 to 250 μL
Unit Size:
Each
Description
- Thumb-friendly soft spring pipetting system reduces fatigue
- 3-year warranty
- Unique side EZIject button option reduces thumb movement
- Finger-hook provides for a more relaxed grip
- Large ergonomic push button is easy on the thumb during contact
- Locking feature prevents accidental volume changes during pipetting procedures
Specifications
Specifications
| Accuracy | ±1.0/ ±0.6% |
| Description | Tip Type: 300 μL |
| Number of Channels | 1 |
| Product Line | Axypet |
| Type | Single-Channel Pipettor |
| Volume (Metric) | 50 to 250 μL |
| Precision | ±0.4/ ±0.3% |
| Warranty | 3 years |