missing translation for 'onlineSavingsMsg'
Learn More
Azelastine hydrochloride, Thermo Scientific Chemicals Product Code.: 16554304

Azelastine hydrochloride, Thermo Scientific Chemicals

Product Code. 16554304
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 16554304

Brand: Thermo Scientific Acros 465080010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79307-93-0
Molecular Formula C22H25Cl2N3O
Molecular Weight (g/mol) 418.36
MDL Number MFCD00242783
InChI Key YEJAJYAHJQIWNU-UHFFFAOYNA-N
IUPAC Name hydrogen 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)-1,2-dihydrophthalazin-1-one chloride
SMILES [H+].[Cl-].CN1CCCC(CC1)N1N=C(CC2=CC=C(Cl)C=C2)C2=CC=CC=C2C1=O

Specifications

Melting Point 225.0°C to 229.0°C
Quantity 1 g
Formula Weight 418.36
Chemical Name or Material Azelastine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0413577 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number465082500,465080010
Lot NumberA0446840 Certificate TypeCertificate of Analysis Date03/11/2022 Catalog Number465082500,465080010
Lot NumberA0413577 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number465082500,465080010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azelastine hydrochloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.