Learn More
Azlon™ Multi-lingual Round Integral Wash Bottle
30.69 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Use promo code "23259" to get your promotional price.
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Description
- Multilingual
- Wide neck wash bottle
- Indelibly printed in English, French, German and Spanish
- Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
- Fitted with color coded one-piece PP closure that maintains it's shape in use
- Designed with integral spout to prevent contamination
Specifications
Capacity (Metric) | 500 mL |
Includes | Integral spout |
Closure Color | White |
Closure Included | Yes |
Mouth | Wide |
Product Type | Wash Bottle |
Chemical Label | Distilled Water |
Closure Material | Polypropylene |