Learn More
Bollé Safety B-Clean Lens Cleaning Products/Kits
Click to view available options
Product Type:
Anti-Fog Spray
Cleaning Station
Lens Cleaner Spray
Multi-function Dry Cleaning Tissues
Tissue Dispenser
Quantity:
1 Ea.
100/Pk.
200/Pk.
250 mL
50 mL
500 mL
Description
- Alcohol and silicone free
- Water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles
Specifications
Specifications
Product Type | Tissue Dispenser |
Quantity | 100/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Product Content Correction
Product Title
Bollé Safety B-Clean Lens Cleaning Products/Kits >