Learn More
Saint Gobain Life Sciences BarbLock™ Hand Tool
BarbLock™ Hand Tool with stainless steel jaw and handle installs BarbLock™ retainers to tubing.
Brand: Saint Gobain Life Sciences BLT135500_OLD
This item is not returnable.
View return policy
Description
BarbLock™ ultra-secure retainers are the most advanced design ideal for systems transferring delicate liquids or gases. They have become the standard in many industries because they solve inherent leak path problems in sensitive and demanding applications. They require no adhesives or solvents and are available in high temperature materials for thermally demanding environments.
- Constructed from high-performance polypropylene that meets USP Class VI criteria
- Quick and easy to assemble
- Compatible with all flexible tubing material
- Disposable assemblies
- Eliminates solvent bonding, ultraviolet bonding and toxic adhesives
- Autoclavable
Specifications
500 Series BarbLock™ Retainers (1/2 in. I.D. tubing) | |
1/2 in. I.D. |