Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories, including a remote dispenser.
1170.00€
Specifications
For Use With (Equipment) Barnstead E-Pure
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11486348
View Documents
Thermo Scientific™
D8952
Remote Dispenser
1170.00€
Each
Estimated Shipment: 23-05-2025
Description
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
Specifications
Specifications
Barnstead E-Pure