Learn More
Emma Safety Footwear BAS XD Safety Shoes
All purpose safety boot. The outsole repels acid and oil like an umbrella repels water. Strong ankle supporting upper fits the foot snugly and comfortably.
Brand: Emma Safety Footwear MM70186843
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
BAS XD | |
Leather | |
43 |
Product Content Correction
Product Title
Emma Safety Footwear BAS XD Safety Shoes > Size: 43