missing translation for 'onlineSavingsMsg'
Learn More

BD Autoclip™ Wound Closing System

Ideal for use on lab animals or in veterinary surgery

610.00€

Specifications

Material Stainless Steel
For Use With (Application) For use on lab animals or in veterinary surgery
Autoclavable Autoclavable
View More Specs

Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
17275043
View Documents
BD
427631
Wound Clip
610.00€
Pack of 1000
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
Description

Description

The Complete Kit (22-275998) consists of 01-804 applier, 01-804-5 nonsterile wound clips, and 01-804-15 remover. To use, insert clips into applier; locate edges of wound and use applier as you would a staple gun to rapidly close wound.

  • Provides maximum holding force with minimal trauma May be used without assistance Easily remove clips with clip remover when wound is healed

Applier

  • Stainless-steel applying forceps hold one rack of 20 clips Clips fed by rust proof beryllium-copper springs Dimensions: 4.5L x 1.37W x 0.37 in.H (114 x 35 x 9.5mm) Net weight: 1.5 oz. (45g)

Wound Clips

  • 9mm nonsterile Autoclip wound clips 20 stainless-steel double clips per rack 100 clips (5 racks) per box, 10 boxes per case

Remover

  • Easily removes clips from healed wound Stainless steel
  • Length: 4 in. (100mm)

Note: Items can be purchased as a kit or individually.

Specifications

Specifications

Stainless Steel
For use on lab animals or in veterinary surgery
Autoclavable
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.