missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Sodium Desoxycholate
Brand: BD 224820
This item is not returnable.
View return policy
Description
- For pneumococcus bile solubility test
- As a media component
Specifications
ISO/DIS, 11014 | |
Sodium Desoxycholate | |
Pneumococcus Bile solubility test and as a media component | |
100 g |
<30°C | |
Dehydrated | |
Bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bottle Sodium Desoxycholate 100G
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H335-May cause respiratory irritation.
Precautionary Statement
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P403-Store in a well-ventilated place.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium 3-roxy-5-ate