missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Nutrient Agar Slants
For cultivation of bacteria and enumeration of organisms in water, sewage, feces and other materials
Brand: BD 220971
This item is not returnable.
View return policy
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, AOAC, BAM, CCAM, COMPF, ISO, SMWW, USDA | |
Powder | |
Nutrient Agar | |
For enumeration of organisms in Water, sewage, feces | |
100 Pack |
Beige | |
United States | |
Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L, Agar 15g/L | |
Tube with Tight Cap |
For in vitro diagnostic use