missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media

Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles

13.15€ - 238.00€

Specifications

Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate (Fluid)
Formulation Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L
View More Specs

Products 2
Product Code Brand Quantity Color Media Type Price Quantity & Availability  
Product Code Brand Quantity Color Media Type Price Quantity & Availability  
12830693
View Documents
BD
221195
10 x 8 mL Light Beige Thioglycollate (Fluid) This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
17372321
View Documents
BD
221196
100 x 8 mL Light Beige Thioglycollate (Fluid)
238.00€
Pack of 100
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
Description

Description

This medium is capable of supporting good growth of a great variety of fastidious organisms, of both pathogenic and nonpathogenic species.

  • Neutralize antibacterial activity of mercurial compounds
  • Before use ∽30% of media should oxidized

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications

Specifications

2°C to 8°C
United States
Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L
Tube
Liquid
Thioglycollate (Fluid)
Sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 183 Days

SDS
Documents

Documents

Product Certifications

For in vitro diagnostic use.