Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefazolin12
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand:  BD 231593
Product Code. 17372381
  • 186.00€ / Pack of 10
Estimated Shipment: 10-12-2024
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cefazolin12
30μg
Disc
10 Pack
Special Offers
For in vitro diagnostic use