Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefpodoxime21
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
186.00€
Specifications
Antimicrobial Agents | Cefpodoxime 21 |
---|---|
Concentration | 10μg |
Packaging Type | Disc |
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Cefpodoxime 21 | |
10μg | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 365 Days
For in vitro diagnostic use