Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefpodoxime21
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
186.00€
Specifications
Antimicrobial Agents Cefpodoxime 21
Concentration 10μg
Packaging Type Disc
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17332401
View Documents Promotion Details
BD
231674
10 Pack
186.00€
Pack of 10
Estimated Shipment: 10-12-2024
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
Cefpodoxime 21
10μg
Disc
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 365 Days
SDS
Documents
Documents
Product Certifications
For in vitro diagnostic use