Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Levofloxacin42
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand: BD 231706
This item is not returnable.
View return policy
Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Levofloxacin42 | |
5μg | |
Disc | |
1 Pack |
For in vitro diagnostic use