Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Ofloxacin56
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Brand:  BD 231672
Product Code. 17342441
  • 189.00€ / Pack of 10
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ofloxacin56
5μg
Disc
10 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For in vitro diagnostic use