Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Bacitracin, B-2
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
Brand:  BD 231267
Product Code. 17342371
  • 188.00€ / Pack of 10
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bacitracin
2U
Disc
10 Cartridges