Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Minocyline
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
51.85€
Specifications
Antimicrobial Agents Minocycline
Concentration 30μg
Packaging Type Disc
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11783819
View Documents Promotion Details
BD
231251
10/Pk.
51.85€
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 02-10-2024
Description
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Minocycline
30μg
Disc
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 1095 Days