Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Minocyline
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
51.85€
Specifications
Antimicrobial Agents | Minocycline |
---|---|
Concentration | 30μg |
Packaging Type | Disc |
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Minocycline | |
30μg | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 1095 Days