Learn More
Brilliant Stain Buffer, BD
Designed to complement multicolor flow cytometry experiments. Brilliant Stain Buffer is a buffer for the immunofluorescent staining of cells.
Brand:  BD Biosciences 563794
142.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15349374
  • 317.00€ / Each
Edge
Description
  • Compatible with the use of other fluorescent staining reagents conjugated with traditional fluorochromes
  • Research Use Only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 Tests
Product Title
Brilliant Stain Buffer, BD > Brilliant Stain buffer; 5mL