Learn More
BD Calcium Assay Kit
For homogeneous measurement of intracellular Calcium changes
Brand: BD 640178
4538.35 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Calcium Indicator, 10X Signal Enhancer
Description
- Assay involves only one dye addition step and does not require washing
- Allows easy implementation in high throughput environment
Specifications
-20°C | |
10 vials calcium indicator,100mL signal enhancer (10X, lyophilized) | |
Detection Assay Kit |
For homogeneous measurement of intracellular calcium changes | |
100 Preps |
Product Content Correction
Product Title
BD Calcium Assay Kit > 1 kit