Learn More
BD Cytometric Bead Array (CBA) Human Angiogenin; Flex Set, Bead C4
For measuring human angiogenin in serum, plasma and cell culture supernatant samples
Brand: BD 558328
Includes: Standard, PE Detection Reagent, Capture Bead C4
Description
- Capture bead and PE detection reagent formulated to 50x concentration
- Includes lyophilized standard
Specifications
Angiogenin | |
1 Vial capture bead 50X, 1 vial detection reagent 50X, 2 vials standard (lyophilized) | |
Serum, Plasma, Cell Culture Supernatant | |
Flow cytometry, for measuring Human angiogenin |
Human | |
100 Tests | |
4°C | |
Human Angiogenin flex set (bead C4) |