Learn More
BD Cytometric Bead Array (CBA) Human Eotaxin Flex Set, Bead C7
Sufficient for 100 tests
Brand: BD 558329
Includes: Standard, PE Detection Reagent, Capture Bead C7
Description
- Capture bead and PE detection reagent formulated to 50x concentration
- Includes lyophilized standard
Specifications
Eotaxin | |
1 Vial capture bead 50X, 1 vial detection reagent 50X, 2 vials standard (lyophilized) | |
Serum, Plasma, Cell Culture Supernatant | |
Flow Cytometry |
Human | |
100 Tests | |
4°C | |
Human eotaxin flex set (bead C7) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Human Eotaxin Flex Set
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT