Learn More
BD Cytometric Bead Array (CBA) Human IL-1β; Flex Set, Bead B4
Bead-based immunoassay capable of measuring human interleukin-1Glc beta (IL-1Glc beta) in serum, plasma, and cell culture supernatant samples
Brand:  BD 558279
Product Code. 15859278
Includes: Human IL-1β; Standard; Human IL-1β; PE Detection Reagent; Human IL-1β; Capture Bead B4
Description
Description
Reactivity
  • Qc Testing: Human
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-1beta
Human IL-1beta, standard, Human IL-1beta, PE detection reagent, Human IL-1beta, capture bead B4
Serum, Plasma, Cell Culture Supernatant
Flow Cytometry
Human
100 Tests
4°C
Human IL-1beta flex set (bead B4)