missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Human IL-12p70 Enhanced Sensitivity Flex Set (Bead C6)

Measures human Interleukin-12p70 (IL-12p70) in serum, plasma and cell culture supernatant samples

Brand:  BD 561518


Product Code. 15821099

  • 740.00€ / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket

Includes:
  • Kit contents:
  • Enhanced Sensitivity Capture Bead C6: stored in aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
  • Detection Reagent (Part A): stored in aqueous buffered solution containing BSA and ProClin™ 150
  • Standard: lyophilized in an aqueous buffered solution containing BSA and ProClin 150
This item is not returnable. View return policy

Description

Description

This kit is used in conjunction with BD™ Cytometric Bead Array (CBA) Human Enhanced Sensitivity Master Buffer Kit, flow cytometer and FCAP Array™ Software and can measue soluble analytes present in very low concentrations.

  • Bead-based immunoassay
  • Reactivity: human
  • Standard assay range: 274 to 200000fg/mL

Flow cytometry, Immunoassay

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lyophilized in an aqueous buffered solution containing BSA™ and ProClin™ 150. Store at 4°C.
Capture beads, detection reagent, standards
Flow Cytometer
Used in conjunction with BD™ cytometric bead array (CBA)
100
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Human IL-12p70 Enhanced Sensitivity Flex Set

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT

SDS
Documents

Documents

Certificates
Special Offers

Special Offers